3-Chloro-2-fluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-2-fluorobenzene-1-sulphonyl chloride, 3-Chloro-2-fluorobenzene-1-sulfonyl chloride

A664620-1G A664620-25G A664620-10G
103840-462EA 18 USD
103840-462 76844-656 76844-658 77158-094
3-Chloro-2-fluorobenzenesulfonyl chloride 98%
3-Chloro-2-fluorobenzenesulphonyl chloride

Caution: for Research Use Only

Formula: C₆H₃Cl₂FO₂S
Boiling Pt: 287.2 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094234
CAS Number: 351003-48-0
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR