2-(3-Bromophenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A271495-1G
A271495-5G
A271495-5G
A271495-1G
A271495-100G
A271495-10G
A271495-25G
103813-048EA
9
USD
103813-048
77276-782
103852-960
77276-780
77305-080
77305-084
77305-082
2-(3-Bromophenyl)ethanol 98%
2-(3-Bromophenyl)ethanol
Caution: for Research Use Only
Formula:
C₈H₉BrO MW: 201.06 g/mol Boiling Pt: 274.6 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191857 CAS Number: 28229-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...