3-Bromo-5-chloropyrazin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-bromo-5-chloropyrazine

A334911-10G A334911-250MG A334911-25G A334911-5G A334911-100G
103825-672EA 8 USD
103825-672 103836-642 76788-228 76788-230 76839-646 76839-648 77872-486
3-Bromo-5-chloropyrazin-2-amine 97%
3-Bromo-5-chloropyrazin-2-amine

Caution: for Research Use Only

Formula: C4H3BrClN3
MW: 208.4400 g/mol
Boiling Pt: 298.4 °C (760 mm Hg)
Melting Pt: 100…104 °C
Flash Pt: 134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909643
CAS Number: 76537-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR