3-Bromo-4,5-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A566892-1G A566892-25G A566892-5G A566892-10G A566892-250MG
103828-780EA 28 USD
103828-780 103839-920 76844-034 76844-036 76844-038 76878-630 76878-632
3-Bromo-4,5-difluorobenzoic acid 97%
3-Bromo-4,5-difluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20233291
CAS Number: 1244642-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR