3-Aminopyrazine-2-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
A350181-1G
A350181-25G
A350181-100MG
A350181-10G
A350181-250MG
A350181-5G
103826-034EA
14
USD
103826-034
103837-028
76840-132
76840-134
77278-144
77278-146
77278-148
77306-608
3-Aminopyrazine-2-carbonitrile 98%
3-Aminopyrazine-2-carbonitrile
Caution: for Research Use Only
Formula:
C5H4N4 Boiling Pt: 364.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD02091510 CAS Number: 25911-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...