3-Aminopiperidine-2,6-dione hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-aminopiperidine-2,6-dione hydrochloride, 2,6-Dioxopiperidine-3-ammonium chloride

A476158-25G A476158-1000G A476158-100G A476158-10G A476158-500G A476158-1G A476158-5G
103828-080EA 5 USD
103828-080 103839-150 76843-078 77034-314 77034-316 77034-318 77034-320 77307-518 77307-520
3-Aminopiperidine-2,6-dione hydrochloride 97%
3-Aminopiperidine-2,6-dione hydrochloride

Caution: for Research Use Only

Formula: C5H9ClN2O2
Storage Temperature: Ambient
MDL Number: MFCD11042437
CAS Number: 24666-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR