3-Aminobenzyl alcohol 95%
Supplier: Ambeed
|
Warning
|
A299579-25G
A299579-100G
A299579-10G
A299579-250MG
A299579-500G
A299579-1G
A299579-5G
103814-232EA
6
USD
103814-232
103835-380
76838-072
77099-480
77099-482
77099-484
77099-486
77305-770
77305-772
3-Aminobenzyl alcohol 95%
3-Aminobenzyl alcohol
Caution: for Research Use Only
Formula:
C₇H₉NO Boiling Pt: 290.7 °C (760 mm Hg) Melting Pt: 92…95 °C Flash Pt: 129.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007817 CAS Number: 1877-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...