3-Amino-6-methoxypyridazine 98%

Supplier: Ambeed
Warning

A231192-1G A231192-25G A231192-5G A231192-10G A231192-250MG
103810-760EA 6 USD
103810-760 103850-508 77089-384 77089-386 77089-388 77275-752 77275-754
3-Amino-6-methoxypyridazine 98%
3-Amino-6-methoxypyridazine

Caution: for Research Use Only

Formula: C₅H₇N₃O
Boiling Pt: 353.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01646127
CAS Number: 7252-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR