3-Amino-2-bromo-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-2-bromo-4-picoline

A267269-100G A267269-10G A267269-250MG A267269-1G A267269-25G A267269-5G
103812-842EA 6 USD
103812-842 103852-742 77001-042 77001-044 77001-046 77092-466 77092-468 77092-470
3-Amino-2-bromo-4-methylpyridine 98%
3-Amino-2-bromo-4-methylpyridine

Caution: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04
Boiling Pt: 308 °C (760 mm Hg)
Flash Pt: 140.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112497
CAS Number: 126325-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR