3,5-Dimethylphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid
A119477-100G
A119477-10G
A119477-25G
A119477-500G
A119477-5G
103830-596EA
8
USD
103830-596
77189-592
77189-594
77189-596
77189-598
77311-890
3,5-Dimethylphenylboronic acid 98%
3,5-Dimethylphenylboronic acid
Caution: for Research Use Only
Formula:
C₈H₁₁BO₂ Boiling Pt: 312.7 °C (760 mm Hg) Melting Pt: 261…265 °C Flash Pt: 142.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185689 CAS Number: 172975-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...