3,5-Dimethoxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A323397-100G
A323397-10G
A323397-500G
A323397-25G
A323397-1G
A323397-5G
103825-338EA
5
USD
103825-338
103836-274
76817-074
76817-076
76817-078
76839-192
77306-294
77306-296
3,5-Dimethoxybenzaldehyde 97%
3,5-Dimethoxybenzaldehyde
Caution: for Research Use Only
Formula:
C₉H₁₀O₃ Boiling Pt: 151 °C (760 mm Hg) Melting Pt: 45…48 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003366 CAS Number: 7311-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...