3,5-Dichlorophenylhydrazinium chloride 97%
Supplier: Ambeed
|
Warning
|
A351216-5G
A351216-100G
A351216-10G
A351216-25G
103837-044EA
12
USD
103837-044
76840-150
77115-110
77115-112
77306-614
3,5-Dichlorophenylhydrazinium chloride 97%
3,5-Dichlorophenylhydrazinium chloride
Caution: for Research Use Only
Formula:
C₆H₇Cl₃N₂ Boiling Pt: 286.1 °C (760 mm Hg) Melting Pt: 208…210 °C Flash Pt: 126.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012938 CAS Number: 63352-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...