3,4,5-Trimethoxyaniline 98%

Supplier: Ambeed
Warning

A177873-5G A177873-500G A177873-5G A177873-25G A177873-10G A177873-100G
77083-008EA 6.37 USD
77083-008 77273-926 103845-610 77083-006 77273-928 77273-930
3,4,5-Trimethoxyaniline 98%
3,4,5-Trimethoxyaniline

Caution: for Research Use Only

Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Boiling Pt: 300.4 °C (760 mm Hg)
Melting Pt: 110…113 °C
Flash Pt: 147.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008393
CAS Number: 24313-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR