2-Hydroxy-3-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)pyridin-2-ol

A172797-25G A172797-100G A172797-10G A172797-500G A172797-5G
103845-042EA 7 USD
103845-042 77082-254 77096-542 77096-544 77096-546 77301-220
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
2-Hydroxy-3-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₆H₄F₃NO
MW: 163.1
Boiling Pt: 223.7 °C (760 mm Hg)
Melting Pt: 150…153 °C
Flash Pt: 89.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00153194
CAS Number: 22245-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR