3-Hydroxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A272295-250MG A272295-1G A272295-25G A272295-5G
103813-090EA 7 USD
103813-090 103853-000 76723-052 77092-792 77305-104 77305-106
3-Hydroxyphenylboronic acid pinacol ester 98%
3-Hydroxyphenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₂H₁₇BO₃
Boiling Pt: 343.5 °C (760 mm Hg)
Melting Pt: 94…98 °C
Flash Pt: 161.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02093755
CAS Number: 214360-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR