2-Nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A135348-10G A135348-5G A135348-100G A135348-1G A135348-500G A135348-1000G A135348-25G A135348-5G
77095-178EA 5.31 USD
77095-178 77298-774 103833-096 77095-172 77095-180 77095-176 77298-772 77095-174
2-Nitrobenzaldehyde 98%
2-Nitrobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 268.2 °C (760 mm Hg)
Melting Pt: 42…44 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR