2-Tolylboronic acid 98%

Supplier: Ambeed
Warning

A203980-25G A203980-100G A203980-10G A203980-500G A203980-5G
103848-174EA 5 USD
103848-174 77086-322 77192-406 77192-408 77192-410 77302-890
2-Tolylboronic acid 98%
2-Tolylboronic acid

Caution: for Research Use Only

Formula: C₇H₉BO₂
Boiling Pt: 283.4 °C (760 mm Hg)
Melting Pt: 162…164 °C
Flash Pt: 125.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00093526
CAS Number: 16419-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR