2-Iodobenzaldehyde 98%

Supplier: Ambeed
Warning

A187888-1G A187888-25G A187888-5G A187888-100G A187888-10G A187888-250MG
103816-048EA 6 USD
103816-048 103846-642 77084-354 77084-356 77084-358 77138-974 77138-976 77138-978
2-Iodobenzaldehyde 98%
2-Iodobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₅IO
Boiling Pt: 266.3 °C (760 mm Hg)
Melting Pt: 36…39 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039570
CAS Number: 26260-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR