2-Iodoaniline 98% (by GC)

Supplier: Ambeed
Warning

A104506-100G A104506-10G A104506-25G A104506-500G A104506-5G
103822-440EA 8 USD
103822-440 77283-220 77283-222 77283-224 77283-226 77283-228
2-Iodoaniline 98% (by GC)
2-Iodoaniline

Caution: for Research Use Only

Formula: C₆H₆IN
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007680
CAS Number: 615-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR