2-Imidazolecarboxylic acid 98%

Supplier: Ambeed
Warning

A141725-10G A141725-250MG A141725-1G A141725-25G A141725-5G
103803-048EA 7 USD
103803-048 103834-084 77109-828 77109-830 77316-222 77316-224 77316-226
2-Imidazolecarboxylic acid 98%
2-Imidazolecarboxylic acid

Caution: for Research Use Only

Formula: C₄H₄N₂O₂
Boiling Pt: 407.9 °C (760 mm Hg)
Melting Pt: 198 °C
Storage Temperature: Freezer
MDL Number: MFCD00270816
CAS Number: 16042-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR