α-Tropolone 97%

Supplier: Ambeed
Warning

A109691-100G A109691-10G A109691-25G A109691-500G A109691-1G A109691-5G
103797-546EA 6 USD
103797-546 103823-352 77093-920 77093-922 77093-924 77093-926 77283-880 77310-156
α-Tropolone 97%
α-Tropolone

Caution: for Research Use Only

Formula: C₇H₆O₂
Boiling Pt: 80…84 °C at 0.075 mmHg
Melting Pt: 50…52 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS Number: 533-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR