2-Formyl-4-thiopheneboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Formylthiophene-4-boronic acid
A202113-1G
A202113-10G
A202113-250MG
103817-358EA
17
USD
103817-358
103847-994
77086-088
77274-882
77274-884
2-Formyl-4-thiopheneboronic acid 97%
2-Formyl-4-thiopheneboronic acid
Caution: for Research Use Only
Formula:
C₅H₅BO₃S Boiling Pt: 395.1 °C (760 mm Hg) Melting Pt: 270…272 °C Flash Pt: 192.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD03002365 CAS Number: 175592-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...