2-Fluoro-6-(trifluoromethyl)benzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A101823-100G
A101823-10G
A101823-25G
A101823-1G
A101823-5G
103796-160EA
6
USD
103796-160
103821-850
77188-566
77188-568
77188-570
77308-744
77308-746
2-Fluoro-6-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-6-(trifluoromethyl)benzaldehyde
Caution: for Research Use Only
Formula:
C₈H₄F₄O Boiling Pt: 189.4 °C (760 mm Hg) Flash Pt: 69.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061273 CAS Number: 60611-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...