2-Ethynylpyridine 98% stabilised

Supplier: Ambeed
Warning


Stabilization: Stabilised with hydroquinone (HQ) 0.1 %

A285740-1G A285740-25G A285740-5G A285740-100G A285740-10G
103813-686EA 8 USD
103813-686 103834-820 76837-358 76837-360 76837-362 77262-716 77262-718
2-Ethynylpyridine 98% stabilised
2-Ethynylpyridine

Caution: for Research Use Only

Formula: C₇H₅N
Boiling Pt: 170.5 °C (760 mm Hg)
Flash Pt: 76.66 °C
Storage Temperature: Freezer
MDL Number: MFCD00041598
CAS Number: 1945-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR