2-Chloro-9H-carbazole 97%

Supplier: Ambeed
Warning

A152479-10G A152479-1G A152479-25G A152479-100G A152479-250MG
103804-398EA 6 USD
103804-398 103842-568 77162-866 77318-186 77318-188 77892-038 77892-039
2-Chloro-9H-carbazole 97%
2-Chloro-9H-carbazole

Caution: Sealed in dry, at room temperature.

Formula: C₁₂H₈ClN
Boiling Pt: 388.7 °C (760 mm Hg)
Melting Pt: 244 °C
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD01444890
CAS Number: 10537-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR