2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4,6-dimethoxy-s-triazine
A159418-1000G
A159418-100G
A159418-500G
A159418-25G
A159418-5G
103805-252EA
5
USD
103805-252
103843-470
77011-140
77011-142
77011-144
77319-354
77319-356
2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine
Caution: for Research Use Only
Formula:
C₅H₆ClN₃O₂ MW: 175.57 g/mol Boiling Pt: 215.1 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00075607 CAS Number: 3140-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...