2-bromo-5-methylthiazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-methylthiazole
A112011-100G
A112011-10G
A112011-250MG
A112011-25G
A112011-1G
A112011-5G
103797-936EA
6
USD
103797-936
103823-764
76781-150
76781-152
76781-154
76781-156
77310-572
77310-574
2-bromo-5-methylthiazole 97%
2-bromo-5-methylthiazole
Caution: for Research Use Only
Formula:
C₄H₄BrNS Boiling Pt: 195 °C (760 mm Hg) Flash Pt: 71.778 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08460610 CAS Number: 41731-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...