2-Bromo-4-nitroimidazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-nitro-1H-imidazole
A470126-5G
A470126-100G
A470126-10G
A470126-1G
A470126-25G
103828-016EA
5
USD
103828-016
103839-090
76843-000
77034-272
77034-274
77307-472
77307-474
2-Bromo-4-nitroimidazole 97%
2-Bromo-4-nitroimidazole
Caution: for Research Use Only
Formula:
C₃H₂BrN₃O₂ Boiling Pt: 402.5 °C (760 mm Hg) Melting Pt: 180…182 °C at N/A mmHg Flash Pt: 197.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD09025308 CAS Number: 65902-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...