2-Aminoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: Quinolin-2-ylamine

A131898-10G A131898-5G A131898-5G A131898-25G A131898-100G A131898-250MG A131898-1G A131898-1G
77314-180EA 171 USD
77314-180 77314-182 103801-652 77599-900 77314-178 103832-560 76688-656 76688-658
2-Aminoquinoline 97%
2-Aminoquinoline

Caution: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 304.9 °C (760 mm Hg)
Melting Pt: 125 °C
Flash Pt: 163.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00038079
CAS Number: 580-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR