2-Aminofluorene 98%

Supplier: Ambeed
Warning

Synonyms: 9H-Fluoren-2-amine, 2-Fluorenamine, Fluoren-2-ylamine

A320528-1G A320528-25G A320528-5G A320528-100G A320528-500G
103825-236EA 5 USD
103825-236 103836-170 76839-048 76839-050 76839-052 77002-848 77002-850
2-Aminofluorene 98%
2-Aminofluorene

Caution: for Research Use Only

Formula: C₁₃H₁₁N
MW: 181.24
Boiling Pt: 327
Melting Pt: 126…131 °C
Density: 1.281
Flash Pt: 188.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001125
CAS Number: 153-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR