2-Amino-6-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-3-fluoroaniline

A101808-100G A101808-25G A101808-5G A101808-10G
103821-840EA 5 USD
103821-840 76765-194 76765-196 77308-738 77623-828
2-Amino-6-fluorobenzonitrile 98%
2-Amino-6-fluorobenzonitrile

Caution: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.13
Boiling Pt: 272
Melting Pt: 127
Flash Pt: 64.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015477
CAS Number: 77326-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR