2-Amino-5-nitropyrimidine 97%

Supplier: Ambeed
Warning

A149181-10G A149181-5G A149181-1G A149181-25G A149181-250MG A149181-100G
103804-012EA 14 USD
103804-012 103842-134 76871-528 77299-712 77317-622 77317-624 77611-380 77883-181
2-Amino-5-nitropyrimidine 97%
2-Amino-5-nitropyrimidine

Caution: for Research Use Only

Formula: C₄H₄N₄O₂
MW: 140.1
Boiling Pt: 402.4 °C (760 mm Hg)
Melting Pt: 235
Density: 1.558
Flash Pt: 197.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00006103
CAS Number: 3073-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR