6-Amino-m-cresol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol
A177881-10G
A177881-5G
A177881-25G
A177881-100G
103845-616EA
6
USD
103845-616
76998-462
77083-014
77301-528
77600-440
6-Amino-m-cresol 98%
6-Amino-m-cresol
Caution: for Research Use Only
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 259 Melting Pt: 162 Flash Pt: 99.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007693 CAS Number: 2835-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...