2-Amino-5-chlorobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloroanthranilonitrile

A392861-25G A392861-5G A392861-100G A392861-10G A392861-500G
103837-950EA 6 USD
103837-950 76841-344 76841-346 77142-762 77142-764 77142-766
2-Amino-5-chlorobenzonitrile 97%
2-Amino-5-chlorobenzonitrile

Caution: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58
Boiling Pt: 101
Melting Pt: 96…99 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017106
CAS Number: 5922-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR