2-Amino-5-bromo-3,4-dimethylpyridine 97%
Supplier: Ambeed
|
Warning
|
A504055-1G
A504055-100MG
A504055-250MG
A504055-10G
103828-318EA
18
USD
103828-318
103839-412
76843-388
77253-342
77253-344
77747-034
2-Amino-5-bromo-3,4-dimethylpyridine 97%
2-Amino-5-bromo-3,4-dimethylpyridine
Caution: for Research Use Only
Formula:
C₇H₉BrN₂ MW: 201.07 g/mol Boiling Pt: 278.2 °C (760 mm Hg) Melting Pt: 133…135 °C Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093947 CAS Number: 374537-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...