2-Amino-5,6-dihydro-4H-benzothiazol-7-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5,6-dihydrobenzo[d]thiazol-7(4H)-one
A106489-25G
A106489-1G
A106489-5G
A106489-250MG
103797-038EA
12
USD
103797-038
103822-792
76780-850
76780-852
77309-646
77309-648
77871-413
2-Amino-5,6-dihydro-4H-benzothiazol-7-one 98%
2-Amino-5,6-dihydro-4H-benzothiazol-7-one
Caution: for Research Use Only
Formula:
C₇H₈N₂OS Boiling Pt: 376 °C (760 mm Hg) Flash Pt: 181.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00460519 CAS Number: 17583-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...