2-Amino-3-bromopyridine 98%

Supplier: Ambeed
Warning

A157025-100G A157025-10G A157025-1G A157025-25G A157025-5G
103804-952EA 5 USD
103804-952 103843-154 77011-016 77011-018 77300-230 77318-964 77318-966
2-Amino-3-bromopyridine 98%
2-Amino-3-bromopyridine

Caution: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.01
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03095187
CAS Number: 13534-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR