2-Amidinopyrimidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A120110-10G
A120110-250MG
A120110-25G
A120110-1G
A120110-100G
103799-886EA
8
USD
103799-886
103830-700
77246-060
77246-062
77246-064
77297-736
77745-884
2-Amidinopyrimidine hydrochloride 97%
2-Amidinopyrimidine hydrochloride
Caution: for Research Use Only
Formula:
C₅H₇ClN₄ MW: 158.5889 g/mol Boiling Pt: 313.1 °C (760 mm Hg) Melting Pt: 180…183 °C Flash Pt: 127.585 °C Storage Temperature: Ambient |
MDL Number:
MFCD02677965 CAS Number: 138588-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...