2-Acetylanthracene 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Anthracenyl)acetone, 2-Anthryl methyl ketone, 1-(2-Anthracenyl)ethanone

A649641-1G A649641-250MG A649641-5G
103840-392EA 246 USD
103840-392 77254-682 77254-684 77308-062
2-Acetylanthracene 97%
2-Acetylanthracene

Caution: for Research Use Only

Formula: C₁₆H₁₂O
MW: 220.27 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00156727
CAS Number: 10210-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR