2,6-Dichloro-4-hydroxypyridine 98%
Supplier: Ambeed
|
Warning
|
A162803-10G
A162803-250MG
A162803-25G
A162803-1G
A162803-5G
103805-658EA
18
USD
103805-658
103843-908
76997-876
76997-878
77300-606
77319-978
77319-980
2,6-Dichloro-4-hydroxypyridine 98%
2,6-Dichloro-4-hydroxypyridine
Caution: for Research Use Only
Formula:
C₅H₃Cl₂NO Melting Pt: 200…204 °C Flash Pt: 100.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00955619 CAS Number: 17228-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...