2,5-Dichloropyrazine 97%

Supplier: Ambeed
Warning

A157402-100G A157402-1G A157402-25G A157402-5G
103805-004EA 14 USD
103805-004 103843-204 77096-042 77319-024 77319-026 77319-028
2,5-Dichloropyrazine 97%
2,5-Dichloropyrazine

Caution: for Research Use Only

Formula: C₄H₂Cl₂N₂
MW: 148.9781 g/mol
Boiling Pt: 184.4 °C (760 mm Hg)
Flash Pt: 81.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04117886
CAS Number: 19745-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR