2,5-Dichloroisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

A434624-25G A434624-100G A434624-5G A434624-1G A434624-10G A434624-1G
103827-580EA 9 USD
103827-580 76877-302 76877-300 76842-364 103838-646 76842-362
2,5-Dichloroisonicotinic acid 98%
2,5-Dichloroisonicotinic acid

Caution: for Research Use Only

Formula: C₆H₃Cl₂NO₂
Melting Pt: 235…240 °C
Flash Pt: 199.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00466640
CAS Number: 88912-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR