2,5-Dibromopyridine 98%

Supplier: Ambeed
Warning

A513026-1000G A513026-100G A513026-10G A513026-500G A513026-25G A513026-5G
103839-524EA 5 USD
103839-524 77279-744 77279-746 77279-748 77279-750 77307-666 77307-668
2,5-Dibromopyridine 98%
2,5-Dibromopyridine

Caution: for Research Use Only

Formula: C₅H₃Br₂N
Melting Pt: 92…95 °C
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006221
CAS Number: 624-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR