2,4,6-Tris(2-pyridyl)-1,3,5-triazine 96%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine

A519171-25G A519171-5G A519171-1G
103828-488EA 7 USD
103828-488 103839-596 76843-600 76843-602 77307-706
2,4,6-Tris(2-pyridyl)-1,3,5-triazine 96%
2,4,6-Tris(2-pyridyl)-1,3,5-triazine

Caution: for Research Use Only

Formula: C₁₈H₁₂N₆
MW: 312.33 g/mol
Boiling Pt: 618.9 °C (760 mm Hg)
Melting Pt: 248…252 °C (760 mm Hg)
Flash Pt: 288.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006045
CAS Number: 3682-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR