Triphenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Triphenylacetic acid

A567510-5G A567510-5G A567510-1G A567510-1G A567510-250MG
76696-270EA 6.18 USD
76696-270 103828-788 76844-048 77307-836 103839-930
Triphenylacetic acid 97%
Triphenylacetic acid

Caution: for Research Use Only

Formula: C₂₀H₁₆O₂
MW: 288.35 g/mol
Boiling Pt: 401.4 °C (760 mm Hg)
Melting Pt: 270…273 °C
Flash Pt: 210.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004185
CAS Number: 595-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR