1,4-Phenylenediacetic acid 97%
Supplier: Ambeed
|
Warning
|
A280934-100G
A280934-10G
A280934-1G
A280934-25G
A280934-5G
103813-526EA
6
USD
103813-526
103853-478
77141-160
77141-162
77141-164
77305-368
77305-370
77305-372
1,4-Phenylenediacetic acid 97%
1,4-Phenylenediacetic acid
Caution: for Research Use Only
Formula:
C₁₀H₁₀O₄ Boiling Pt: 410.7 °C (760 mm Hg) Melting Pt: 250…256 °C Flash Pt: 216.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004354 CAS Number: 7325-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...