2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: DavePhos2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl

A159031-1G A159031-5G A159031-1G A159031-100G A159031-5G A159031-10G A159031-25G A159031-250MG
77319-292EA 8.24 USD
77319-292 77319-294 77319-296 103805-216 76997-696 76997-694 103843-428 76997-692
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl

Caution: for Research Use Only

Formula: C₂₆H₃₆NP
MW: 393.55 g/mol
Boiling Pt: 539.6 °C (760 mm Hg)
Melting Pt: 121…124 °C
Flash Pt: 280.1 °C
Storage Temperature: Ambient
MDL Number: MFCD02183572
CAS Number: 213697-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR