2-Acetamido-6-nitrobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Acetamido-6-nitrobenzoic Acid, 2'-Carboxy-3'-nitroacetanilide, N-Acetyl-6-nitroanthranilic acid, 2-(Acetylamino)-6-nitrobenzoic Acid, N-Acetyl-6-nitroanthranilic Acid
A703476-5G
A703476-250MG
103840-656EA
57
USD
103840-656
76844-870
77267-602
2-Acetamido-6-nitrobenzoic acid 98%
2-Acetamido-6-nitrobenzoic acid
Caution: for Research Use Only
Formula:
C₉H₈N₂O₅ Boiling Pt: 472.2 °C (760 mm Hg) Flash Pt: 239.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276267 CAS Number: 73721-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...