2-(4-Chlorophenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophenethyl alcohol

A238738-25G A238738-1000G A238738-100G A238738-10G A238738-500G A238738-5G
103851-034EA 5 USD
103851-034 77090-168 77288-688 77288-690 77288-692 77288-694 77304-118
2-(4-Chlorophenyl)ethanol 98%
2-(4-Chlorophenyl)ethanol

Caution: for Research Use Only

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 259
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002899
CAS Number: 1875-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR