2-(4-Aminobenzyl)aniline 95%

Supplier: Ambeed
Danger

Synonyms: 2-[(4-aminophenyl)methyl]aniline

A256248-1G A256248-100MG A256248-250MG
103792-504EA 130 USD
103792-504 103812-254 77091-606 77250-036 77250-038
2-(4-Aminobenzyl)aniline 95%
2-(4-Aminobenzyl)aniline

Caution: for Research Use Only

Formula: C₁₃H₁₄N₂
Boiling Pt: 389.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01670227
CAS Number: 1208-52-2
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR